Please login to your online account to display your discounted pricing

Oxalyl chloride, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  129611000

 View more versions of this product

Catalog No. AC129611000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 126.93
Quantity 100g
Color Clear colorless to pale yellow
Beilstein 02, 542
Merck Index 15, 7013
EINECS Number 201-200-2
TSCA TSCA
RTECS Number KI2950000
Density 1.4780g/mL
Fieser 01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
Refractive Index 1.4285 to 1.4305 (20°C, 589nm)
CAS 76-02-8
CAS 75-44-5
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym Ethanedioyl dichloride
Health Hazard 2 GHS P Statement
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Chemical Name or Material Oxalyl chloride
Assay Percent Range 98%
Linear Formula ClCOCOCl
MDL Number MFCD00000704
CAS 79-37-8
Molecular Formula C2Cl2O2

Specifications

Chemical Name or Material Oxalyl chloride
CAS 79-37-8
CAS 75-44-5
CAS 76-02-8
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2Cl2O2
Linear Formula ClCOCOCl
Formula Weight 126.93
MDL Number MFCD00000704
Synonym Ethanedioyl dichloride
Accessories

Danger

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents