Please login to your online account to display your discounted pricing

Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC369188000

 View more versions of this product

Catalog No. AC369188000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 126.93
Quantity 800mL
Color Clear colorless to yellow
Packaging AcroSeal™ Glass Bottle
Density 1.3350g/mL
Beilstein 02, 542
Fieser 01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
Merck Index 15, 7013
Linear Formula ClCOCOCl
CAS 75-09-2
Chemical Name or Material Oxalyl chloride
Name Note 2.0M solution in dichloromethane
Molecular Formula C2Cl2O2
MDL Number MFCD00000704
CAS 79-37-8
Synonym Ethanedioyl dichloride
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Reacts violently with water.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C

Specifications

Chemical Name or Material Oxalyl chloride
Name Note 2.0M solution in dichloromethane
CAS 79-37-8
CAS 75-09-2
Molecular Formula C2Cl2O2
Linear Formula ClCOCOCl
Formula Weight 126.93
MDL Number MFCD00000704
Synonym Ethanedioyl dichloride
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Reacts violently with water.
Contact with water liberates toxic gas.

Refrigerator +4°C

SDS & Safety Documents