Please login to your online account to display your discounted pricing

Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  369181000*

 View more versions of this product

Catalog No. AC369181000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 126.93
Physical Form Liquid
Quantity 100mL
Color Clear colorless to yellow
Merck Index 15, 7013
Beilstein 02, 542
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 1.3350g/mL
Fieser 01,767; 02,301; 03,216; 04,361; 05,481; 06,424; 07,257; 08,366; 11,379; 14,150; 16,149; 17,241
Linear Formula ClCOCOCl
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 2.0M solution in dichloromethane
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Reacts violently with water.
Contact with water liberates toxic gas.
CAS 75-09-2
CAS 79-37-8
Health Hazard 1 Danger
Synonym Ethanedioyl dichloride
Chemical Name or Material Oxalyl chloride
Molecular Formula C2Cl2O2
Recommended Storage Refrigerator +4°C
MDL Number MFCD00000704

Specifications

Chemical Name or Material Oxalyl chloride
Name Note 2.0M solution in dichloromethane
CAS 79-37-8
CAS 75-09-2
Molecular Formula C2Cl2O2
Linear Formula ClCOCOCl
Formula Weight 126.93
MDL Number MFCD00000704
Synonym Ethanedioyl dichloride
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Reacts violently with water.
Contact with water liberates toxic gas.

Refrigerator +4°C

SDS & Safety Documents