Please login to your online account to display your discounted pricing

Oxalyl bromide, 98%, ACROS Organics™

$210.56 - $210.56

Chemical Identifiers

Chemical Name or Material Oxalyl bromide
Assay ≥97.5% (Argentometry)
Assay Percent Range 98%
Molecular Formula C2Br2O2
Linear Formula BrCOCOBr
Formula Weight 215.83
MDL Number MFCD00000113
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC150895000

  SDS
thermo fisher scientific
AC150895000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC150890250

  SDS
thermo fisher scientific
150890250
Glass bottle 25g Each for $210.56

AC150891000

  SDS
thermo fisher scientific
AC150891000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5% (Argentometry)
Formula Weight 215.83
Quantity 500g
Color Clear
Fieser 01,767; 01,1137
Density 2.3000g/mL
Chemical Name or Material Oxalyl bromide
Assay Percent Range 98%
Molecular Formula C2Br2O2
CAS 15219-34-8
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula BrCOCOBr
MDL Number MFCD00000113
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.5200 to 1.5230 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

C-X bond formation reagent, Oxidation reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.
Reacts violently with water.