Please login to your online account to display your discounted pricing

Alfa Aesar™ Oxalic dihydrazide, 98%

$155.20 - $690.09

Chemical Identifiers

Chemical Name or Material Oxalic dihydrazide
CAS 996-98-5
Molecular Formula C2H6N4O2
Formula Weight 118.1
MDL Number MFCD00007608
Beilstein 1072110
Synonym Oxalyl dihydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1515122

  SDS
alfa aesar™
A1515122
100g Each for $155.20

AAA1515136

  SDS
alfa aesar™
A1515136
500g Each for $690.09
Description & Specifications

Specifications

Formula Weight 118.1
Melting Point ≈240°C (dec.)
TSCA TSCA
CAS 996-98-5
Beilstein 1072110
Quantity 100g
Chemical Name or Material Oxalic dihydrazide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym Oxalyl dihydrazide
RTECS Number RO2840000
Molecular Formula C2H6N4O2
Assay 98%
Health Hazard 1 Warning
MDL Number MFCD00007608
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.