missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Oxalic dihydrazide, 98%

Manufacturer:  Alfa Aesar™ A1515122

 View more versions of this product

Catalog No. AAA1515122



Specifications

Specifications

Oxalic dihydrazide
100g
C2H6N4O2
1072110
SWRGUMCEJHQWEE-UHFFFAOYSA-N
ethanedihydrazide
13826
98%
~240°C (decomposition)
996-98-5
MFCD00007608
oxalyl dihydrazide, oxalic dihydrazide, oxaldihydrazide, ethanedioic acid, dihydrazide, oxalyldihydrazide, oxalhydrazide, oxalylhydrazine, oxaloylhydrazine, oxalic hydrazide, oxalyl hydrazide
C(=O)(C(=O)NN)NN
118.096
118.1

Chemical Identifiers

996-98-5
118.096
SWRGUMCEJHQWEE-UHFFFAOYSA-N
13826
C(=O)(C(=O)NN)NN
C2H6N4O2
MFCD00007608
oxalyl dihydrazide, oxalic dihydrazide, oxaldihydrazide, ethanedioic acid, dihydrazide, oxalyldihydrazide, oxalhydrazide, oxalylhydrazine, oxaloylhydrazine, oxalic hydrazide, oxalyl hydrazide
ethanedihydrazide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RO2840000

TSCA : TSCA

Documents