Please login to your online account to display your discounted pricing

Oxalic acid, potassium salt dihydrate, 99%, extra pure, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC352110010

 View more versions of this product

Catalog No. AC352110010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (Acid)
Purity Grade Extra Pure
Physical Form Crystalline Powder or Solid
Formula Weight 254.2
Quantity 1kg
Color White to beige
Merck Index 15, 7806
Synonym Potassium oxalate (2:1) dihydrate
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Water 16% max. (K.F.)
Name Note 99%
Assay Percent Range 99%
Infrared Spectrum Authentic
MDL Number MFCD00150443
CAS 6100-20-5
Loss on Drying 12 % to 16 % (1 g, 105°C)
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Chemical Name or Material Oxalic acid, potassium salt dihydrate
Molecular Formula C4H3KO8·2H2O
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Oxalic acid, potassium salt dihydrate
Name Note 99%
CAS 6100-20-5
Purity Grade Extra Pure
Assay ≥98.5% (Acid)
Assay Percent Range 99%
Molecular Formula C4H3KO8·2H2O
Formula Weight 254.2
MDL Number MFCD00150443
Synonym Potassium oxalate (2:1) dihydrate
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

SDS & Safety Documents