Please login to your online account to display your discounted pricing

Oxalic Acid Dihydrate (Crystalline/Technical), Fisher Chemical

$164.53 - $647.60

Chemical Identifiers

Chemical Name or Material Oxalic Acid Dihydrate
CAS 6153-56-6
Purity Grade Technical Grade
Physical Form Powder/Solid
Molecular Formula C2H2O4 · 2 H2O
Linear Formula HO2CCO2H·2H2O
Formula Weight 126.07
MDL Number MFCD00149102
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A218-3

  SDS
fisher scientific company
FLA2183
Poly Bottle 3kg Each for $647.60
Case of 4 Each for $2,590.40

A218-500

  SDS
fisher scientific company
FLA218500
Poly Bottle 500g Each for $164.53
Case of 6 Each for $987.18
Description & Specifications

Specifications

Melting Point 98°C
Purity Grade Technical Grade
Physical Form Powder/Solid
Formula Weight 126.07
Quantity 3kg
Density 4.62
DOT Information DOT Class 8, : Corrosive
pH 1.3
Color White
Molecular Formula C2H2O4 · 2 H2O
Vapor Pressure 21.5mbar at 50°C
Identification Pass Test
Health Hazard 3 Emergency Overview
Causes severe eye burns. Harmful in contact with skin and if swallowed. Irritating to respiratory system. Corrosive to metals. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:0
Additional Information Vapor Pressure: 21.5mbar at 50°C
MDL Number MFCD00149102
ChemAlert Storage Symbol White
UN Number 3261
Health Hazard 2 DANGER!
Packaging Poly Bottle
Chemical Name or Material Oxalic Acid Dihydrate
CAS 6153-56-6
Linear Formula HO2CCO2H·2H2O
Safety and Handling
 

Emergency Overview
Causes severe eye burns. Harmful in contact with skin and if swallowed. Irritating to respiratory system. Corrosive to metals. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:0

DANGER!