Please login to your online account to display your discounted pricing

Oxalic Acid, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$185.61 - $1,288.44

Chemical Identifiers

Chemical Name or Material Oxalic Acid
CAS 6153-56-6
Name Note Dihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula HOCOCOOH·2H2O
Formula Weight 126.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003513

 
avantor performance
023001
Poly Bottle 500g Each for $185.61

02003512

 
avantor performance
023005
Poly Bottle 2.5kg Each for $1,288.44
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 126.07
Molecular Formula HOCOCOOH·2H2O
CAS 6153-56-6
Chemical Name or Material Oxalic Acid
Specific Gravity 1L = 1.65kg
Quantity 500g
Name Note Dihydrate
Packaging Poly Bottle