Please login to your online account to display your discounted pricing

Oxalic Acid , Dihydrate, ACS Grade, 99.5 to 102.05%, LabChem™

$167.51 - $167.51

Chemical Identifiers

Chemical Name or Material Oxalic Acid
CAS 6153-56-6
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula HO2CCO2H
Formula Weight 90.04
UN Number UN3261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC180401

 
lab chem
LC180401
500g Each for $167.51
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 90.04
CAS Max % 100
Chemical Name or Material Oxalic Acid
Specific Gravity 1653kg/m3
Solubility Soluble in water
EINECS Number 205-634-3
Decomposition Information Carbon monoxide; Carbon dioxide
Melting Point 101°C
Color White
Quantity 500g
Name Note Dihydrate
Purity Grade ACS Grade
Assay 99.5 to 102.05%
Assay Percent Notes as dihydrate
Linear Formula HO2CCO2H
CAS 6153-56-6
UN Number UN3261
Density 1,653kg/m3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Identification Passes Test
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
Recommended Storage Room Temperature
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.

Room Temperature