Learn More
Oxalic acid dihydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423152500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Oxalic acid dihydrate | |
98.0°C to 102.0°C | |
White | |
C2H6O6 | |
MFCD00149102 | |
02, 502 | |
15, 7010 | |
Solubility in water: 138g/L (20°C). Other solubilities: 1200g/L in water (90°C), 400g/L in ethanol, 555g/L in boiling alcohol, 181g/L in glycerol, 10g/L in ether | |
C(=O)(C(=O)O)O.O.O | |
126.064 | |
126.04 | |
ACS Reagent | |
250 g | |
Plastic bottle | |
0.01% max. |
For analysis ACS | |
6153-56-6 | |
99.5+% | |
HO2CCO2H·2H2O | |
3261 | |
01,764; 02,301; 05,481 | |
oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
oxalic acid;dihydrate | |
61373 | |
≥99.5% | |
4.62 | |
Authentic | |
Fine Crystalline Powder | |
21.5mbar at 50°C |
Chemical Identifiers
6153-56-6 | |
126.064 | |
GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
61373 | |
C(=O)(C(=O)O)O.O.O |
C2H6O6 | |
MFCD00149102 | |
oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
oxalic acid;dihydrate |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only