Learn More
Oxalic acid, 98%, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 186430050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Oxalic acid | |
144-62-7 | |
98% | |
MFCD00002573 | |
15, 7010 | |
Solubility in water: 90g/L (20°C). Other solubilities: very soluble in ethanol, soluble in glycerol, slightly soluble in ether, insoluble in benzene, chloroform and petroleum, ether | |
C(=O)(C(=O)O)O | |
90.034 | |
CHEBI:16995 | |
98% | |
5 kg | |
Authentic | |
Crystalline Powder |
189.0°C to 191.0°C | |
White | |
C2H2O4 | |
02, 502 | |
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
oxalic acid | |
971 | |
90.04 | |
1.9000g/mL | |
>93.4°C | |
Plastic drum | |
1.9 |
Chemical Identifiers
144-62-7 | |
90.034 | |
MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
971 | |
oxalic acid |
C2H2O4 | |
MFCD00002573 | |
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
CHEBI:16995 | |
C(=O)(C(=O)O)O |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only