Please login to your online account to display your discounted pricing

Oxalic Acid, 7.5% (w/v) Aqueous Solution, APHA for Silica, Ricca Chemical

$59.09 - $59.09

Chemical Identifiers

Chemical Name or Material Oxalic Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5467-16

 
ricca chemical company
RI546716
Plastic 500mL Each for $59.09

5467-1

 
ricca chemical company
RI54671
Plastic 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5467-32

 
ricca chemical company
RI546732
Plastic 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Methods APHA for Silica
Chemical Name or Material Oxalic Acid
Concentration or Composition (by Analyte or Components) 7.5% (w/v) Aqueous Solution
Packaging Plastic