missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxalacetic acid, 98+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1578904

 View more versions of this product

Catalog No. AA1578904



Specifications

Specifications

Oxalacetic acid
328-42-7
C4H4O5
UN3261
14,6909
KHPXUQMNIQBQEV-UHFFFAOYSA-N
2-oxobutanedioic acid
970
132.07
≥98%
2g
>98%
MFCD00002592
1705475
oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate
C(C(=O)C(=O)O)C(=O)O
132.071
CHEBI:30744
Powder

Chemical Identifiers

328-42-7
132.071
KHPXUQMNIQBQEV-UHFFFAOYSA-N
970
2-oxobutanedioic acid
C4H4O5
MFCD00002592
oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate
CHEBI:30744
C(C(=O)C(=O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318

GHS P Statement
P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P363-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents