missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Oxacillin sodium salt monohydrate, 815μg/mg

$52.80 - $52.80

Chemical Identifiers

CAS 7240-38-2
Formula Weight 441.43
MDL Number MFCD00167146
Beilstein 4287093
Merck Index 14,6904
Chemical Name or Material Oxacillin sodium salt monohydrate, 815μg/mg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6638003 SDS Alfa Aesar™
J6638003
1g Each for $52.80
Specifications

Description

  • Used primarily against gram positive organisms namely resistant Staphylococcus species
Description & Specifications

Chemical Identifiers

7240-38-2
MFCD00167146
14,6904
441.43
4287093
Oxacillin sodium salt monohydrate, 815μg/mg

Specifications

7240-38-2
441.43
4287093
Freely soluble in water
White
1g
Powder
MFCD00167146
14,6904
687°C
188°C
Oxacillin sodium salt monohydrate, 815μg/mg

Safety and Handling

storageNote1 : 2° to 8°C