Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty one-piece combs, for use with the Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System.
$128.71 - $134.76
Specifications
For Use With (Equipment) Owl™ EasyCast B1 Mini Gel Electrophoresis System
Gel Size Mini
Product Line Owl, EasyCast
Type Comb
View More Specs
Products 3
Catalog Number Mfr. No. Description Teeth Count Thickness (Metric) Teeth Price Quantity  
Catalog Number Mfr. No. Description Teeth Count Thickness (Metric) Teeth Price Quantity  
OW-B1-PREP
View Documents Promotion Details
Thermo Scientific™
B1PREP
Prep comb; 1.5mm thick 2 1.5 mm
Each for $134.76
OWB1RL17D
View Documents Promotion Details
Thermo Scientific™
B1RL17D
Rapid-load 17 tooth (2X) comb; 1.5mm 17 (2X) 1.5 mm
Each for $128.71
OWB1RL9D
View Documents Promotion Details
Thermo Scientific™
B1RL9D
Rapid-load 9 tooth (1X) comb; 1.5mm 9 (1X) 1.5 mm
Each for $134.76
Description
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs. These heavy-duty one-piece combs are for use with the EasyCast™ B1 Mini Gel Electrophoresis System.
Ordering Information:
  • Cat Nos. B1-Rl-9C, -9D, -17C and -17D are 8 and 12-channel pipette format (1X) and (2X) combs
  • Customized combs are available (Order SKU # XCM); please contact your customer service or sales representative for further details
Specifications
Specifications
Owl™ EasyCast B1 Mini Gel Electrophoresis System
Mini
Owl, EasyCast
Comb
SDS
missing translation for 'documents'
missing translation for 'documents'