missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System

Obtain flat, even banding patterns while conserving valuable bench space with the Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System.

$1,475.49 - $1,475.49

Specifications

Certifications/Compliance CE marked
Dimensions (L x W x H) 9.1 x 10.6 x 5.5 in. (23 x 27 x 14 cm)
Height (English) 5.5 in.
Height (Metric) 14 cm
Length (English) 9.1 in.
View More Specs

Products 1
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
09-528-205
View Documents
Thermo Scientific™
D4
Complete system
Each for $1,475.49
 
Description

Description

Obtain flat, even banding patterns while conserving valuable bench space with the Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System.

Features:
  • Designed to provide flat, even banding patterns and consistent results with no tape, grease, agarose seals or other accessories.
  • Can run one gel tray (51 samples) in the buffer chamber, or to save bench space, two stacked trays (102 samples).
  • Two 16 x 17cm UV Transmissible (UVT) gel trays accommodate up to three combs each, allowing the user to run up to three series of samples equal distances.
  • Stand-alone casting platform included for casting two gels simultaneously.
  • A single gel can be cast right in the buffer chamber or in the external caster.

Includes:

  • Buffer chamber (buffer volume, 1.8L)
  • Three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Two UVT gel trays
  • Six 1.5mm thick combs
  • External gel caster
  • Bubble level
Specifications

Specifications

CE marked
5.5 in.
9.1 in.
17 cm
1.8 L
6.5
16 cm
9.1 x 10.6 x 5.5 in. (23 x 27 x 14 cm)
14 cm
23 cm
17 to 102
10.6 in.
27 cm
SDS
Product Certifications