missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System

Obtain flat, even banding patterns while conserving valuable bench space with the Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System.

Manufacturer:  Thermo Scientific™ D4

 View more versions of this product

Catalog No. 09-528-205



Description

Description

Obtain flat, even banding patterns while conserving valuable bench space with the Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System.

Features:
  • Designed to provide flat, even banding patterns and consistent results with no tape, grease, agarose seals or other accessories.
  • Can run one gel tray (51 samples) in the buffer chamber, or to save bench space, two stacked trays (102 samples).
  • Two 16 x 17cm UV Transmissible (UVT) gel trays accommodate up to three combs each, allowing the user to run up to three series of samples equal distances.
  • Stand-alone casting platform included for casting two gels simultaneously.
  • A single gel can be cast right in the buffer chamber or in the external caster.

Includes:

  • Buffer chamber (buffer volume, 1.8L)
  • Three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Two UVT gel trays
  • Six 1.5mm thick combs
  • External gel caster
  • Bubble level
Specifications

Specifications

Owl™
Complete system
Space Saving
14 cm
23 cm
17 to 102
10.6 in.
27 cm
CE marked
9.1 x 10.6 x 5.5 in. (23 x 27 x 14 cm)
5.5 in.
9.1 in.
17 cm
1.8 L
6.5
16 cm
SDS
Documents
Product Certifications