Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Combs
Eliminate comb assembly with heavy-duty, one-piece microwell Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Combs.
$109.44 - $109.44
Specifications
Description Micro well comb
For Use With Thermo Scientific™ Owl D4 Electrophoresis System
Teeth Count 17
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Teeth Count Thickness (Metric) Teeth Price Quantity  
Catalog Number Mfr. No. Description Teeth Count Thickness (Metric) Teeth Price Quantity  
09-528-210 Thermo Scientific™
D417C
Micro well comb 17 1mm Each for $109.44
Specifications
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Combs. These heavy-duty, one-piece microwell combs are for use with the Owl D4 Electrophoresis System.
Ordering Information:
Customized combs are available (Order SKU # XCM). Please contact your customer service or sales representative for further details.
Description & Specifications
Specifications
Micro well comb
Thermo Scientific™ Owl D4 Electrophoresis System
17