Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
$42.22 - $320.34
Specifications
For Use With (Equipment) | Owl™ D2 Wide Gel Electrophoresis System |
---|---|
Product Line | Owl |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
OW-D2CST
|
Thermo Scientific™
D2CST |
External caster (no trays/combs) |
Each for $277.66
|
|
|||||
OW-D2GK
|
Thermo Scientific™
D2GK |
Replacement Gaskets (1 pair) |
Pack of 2 Each for $42.22
|
|
|||||
09-528-166
|
Thermo Scientific™
D2UVT |
EasyCast™ gasketed UVT gel tray |
Each for $320.34
|
|
|||||
Description
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
- UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
- Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
Specifications
Owl™ D2 Wide Gel Electrophoresis System | |
Owl |