missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Owl™ A6 Wide Gel Horizontal Electrophoresis System

Produce clear, tight banding patterns with the Thermo Scientific™ Owl™ A6 Wide Gel Horizontal Electrophoresis System, an ideal system for high-throughput laboratories.

Manufacturer:  Thermo Scientific™ A6

Catalog No. OWA6



Description

Description

Produce clear, tight banding patterns with the Thermo Scientific™ Owl™ A6 Wide Gel Horizontal Electrophoresis System, an ideal system for high-throughput laboratories.

Features:
  • Screens 25 to 500 samples on a single agarose gel, producing clear, tight banding patterns with no “smiling.”
  • Unique comb-slot design of the gel tray allows use of up to 10 evenly spaced combs.
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96, eight or 12-well plate.
  • Incorporates Owl's end-gate casting design—gel tray is fitted with gasketed end gates which provide a leakproof seal without tape.
  • Sample loading with multichannel pipetter speeds up transfer.
  • Bubble level ensures casting of flat, uniformly thick gels.

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray with gasketed end gates
  • Four combs (two 25-well and two 50-well, 1.5mm thick)
Specifications

Specifications

23 cm x 25 cm
Owl™
Gel size: 23 x 25cm; Buffer volume: 3L; L x W x H: 35 x 38 x 11cm
Wide
11 cm
35 cm
Model A6
3 L
38 cm
250 V
CE marked
13.75 x 14.94 x 4.38 in. (35 x 38 x 11 cm)
4.38 in.
13.75 in.
25 cm
25 to 500
14.94 in.
23 cm
SDS
Documents
Product Certifications
Promotions

Promotions