Please login to your online account to display your discounted pricing

Thermo Scientific™ Owl™ A1 Large Gel System

Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems, which have smaller footprints ideal for detailed RNA/DNA analysis.

$1,333.96 - $1,584.37

Specifications

Certifications/Compliance CE marked
Dimensions (L x W x H) Exterior 14.8 x 8.5 x 4.3 in. (37.5 x 21.5 x 11cm)
Height (English) Exterior 4.3 in.
Height (Metric) Exterior 11cm
Length (English) Exterior 14.8 in.
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    

09-528-100

 
thermo scientific™
A1*
Large Each for $1,333.96

09-528-100A

 
thermo scientific™
A1BP
Large, w/buffer exchange ports Each for $1,584.37
Description & Specifications

Specifications

Certifications/Compliance CE marked
Dimensions (L x W x H) Exterior 14.8 x 8.5 x 4.3 in. (37.5 x 21.5 x 11cm)
Height (English) Exterior 4.3 in.
Height (Metric) Exterior 11cm
Length (English) Exterior 14.8 in.
Length (Metric) Exterior 37.5cm
Length (Metric) Gel 25cm
Sample Range 8 to 150
Volume (Metric) Buffer 1.6L
Width (English) Exterior 8.5 in.
Width (Metric) Exterior 21.5cm
Width (Metric) Gel 13cm

Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems. With one of the smallest footprints of any large-format device, these systems are ideal for detailed RNA/DNA analysis.

Features:

  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • Programmable power inverter may be used for field reversal electrophoresis
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (12-, 16- and 20-well), 1.5mm thick