Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts
Choose from various Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts, including gasketed UVT gel trays, replacement gaskets and external casters.
Manufacturer:  Thermo Scientific™ A1UVT
 View more versions of this product
Catalog No. OW-A1UVT
Description
Description
Choose from various Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts, including gasketed UVT gel trays, replacement gaskets and external casters.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 10 sample sets on the same gel
  • Unique electrode placement allows this gel to be run quickly without compromising the resolution
Specifications
Specifications
UVT tray with gasketed end gates
Owl™ A1 Large Gel Electrophoresis System
Gel Tray