Please login to your online account to display your discounted pricing

OVA257-264 (SIINFEKL) Mouse anti-Mouse, PerCP-eFluor 710, Clone: eBio25-D1.16 (25-D1.16), eBioscience™

Mouse Monoclonal Antibody

$158.05 - $477.05

Specifications

Antigen OVA257-264 (SIINFEKL)
Clone eBio25-D1.16 (25-D1.16)
Host Species Murine
Gene Alias H-2Kb-SIINFEKL, OVA-Kb
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016069

 
life technologies
LS46574380
25μg Each for $158.05

5016070

 
life technologies
LS46574382
100μg Each for $477.05
Description & Specifications

Specifications

Antigen OVA257-264 (SIINFEKL)
Clone eBio25-D1.16 (25-D1.16)
Host Species Murine
Gene Alias H-2Kb-SIINFEKL, OVA-Kb
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 25-D1.16 monoclonal antibody reacts with the ovalbumin-derived peptide SIINFEKL bound to H-2Kb of MHC class I, but not with unbound H-2Kb, or H-2Kb bound with an irrelevant peptide. This antibody has proven to be very useful tracking the quantity and localization of these specific antigen-presenting cells (APC) in vivo.