missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OVA257-264 (SIINFEKL) Mouse anti-Mouse, PE, Clone: eBio25-D1.16 (25-D1.16), eBioscience™

Mouse Monoclonal Antibody

$223.08 - $460.68

Specifications

Antigen OVA257-264 (SIINFEKL)
Clone eBio25-D1.16 (25-D1.16)
Host Species Murine
Gene Alias H-2Kb-SIINFEKL, OVA-Kb
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010844 Life Technologies
LS12574381
50μg Each for $223.08
5010845 Life Technologies
LS12574382
100μg Each for $460.68
Specifications

Description

The 25-D1.16 monoclonal antibody reacts with the ovalbumin-derived peptide SIINFEKL bound to H-2Kb of MHC class I, but not with unbound H-2Kb, or H-2Kb bound with an irrelevant peptide. This antibody has proven to be very useful tracking the quantity and localization of these specific antigen-presenting cells (APC) in vivo.
Description & Specifications

Specifications

OVA257-264 (SIINFEKL)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
eBio25-D1.16 (25-D1.16)
H-2Kb-SIINFEKL, OVA-Kb
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer