missing translation for 'onlineSavingsMsg'
Learn More

Osthole 98.0+%, TCI America™

Supplier:  TCI America O04261G

 View more versions of this product

Catalog No. O04261G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Osthole
84°C
C15H16O3
1 g
MBRLOUHOWLUMFF-UHFFFAOYSA-N
7-methoxy-8-(3-methylbut-2-enyl)chromen-2-one
10228
244.29
Crystalline Powder
484-12-8
White-Yellow
MFCD00076049
7-Methoxy-8-(3-methyl-2-butenyl)coumarin
CC(=CCC1=C(C=CC2=C1OC(=O)C=C2)OC)C
244.29
CHEBI:69832
≥98.0% (GC)

Chemical Identifiers

484-12-8
244.29
MBRLOUHOWLUMFF-UHFFFAOYSA-N
10228
7-methoxy-8-(3-methylbut-2-enyl)chromen-2-one
C15H16O3
MFCD00076049
7-Methoxy-8-(3-methyl-2-butenyl)coumarin
CHEBI:69832
CC(=CCC1=C(C=CC2=C1OC(=O)C=C2)OC)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GN7700000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions