Please login to your online account to display your discounted pricing

Osmium(VIII) oxide, For Microscopy, EMD Millipore™


Manufacturer: emd millipore corporation  1245050100

 View more versions of this product

Catalog No. M1245050100

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 254.19
DOT Information DOT Class: 6.1, Packaging Group: I
Chemical Name or Material Osmium(VIII) oxide
EINECS Number 244-058-7
Odor Stinging
Packaging Glass ampoule
Physical Form Solid
Molecular Formula OsO4
Appearance Yellow solid
CAS 20816-12-0
UN Number UN 2471
Vapor Pressure 10 hPa at 20°C
Synonym Osmic acid
Health Hazard 1 H300 + H310 + H330: Fatal if swallowed, in contact with skin or if inhaled
H314: Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material Osmium(VIII) oxide
CAS 20816-12-0
Molecular Formula OsO4
Formula Weight 254.19
UN Number UN 2471
Synonym Osmic acid
Accessories

H300 + H310 + H330: Fatal if swallowed, in contact with skin or if inhaled
H314: Causes severe skin burns and eye damage.