missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Osmium(VIII) oxide, For Microscopy, EMD Millipore™

$329.97 - $1,098.03

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.226
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1245050001 View Documents Millipore Sigma
1245050001
1g Glass ampoule Each for $1,098.03
M1245050100 View Documents Millipore Sigma
1245050100
100mg Glass ampoule Each for $329.97
M1245050500 View Documents Millipore Sigma
1245050500
500mg Glass ampoule Each for $659.89
Description

Description

For microscopy
Specifications

Chemical Identifiers

20816-12-0
254.226
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
tetraoxoosmium
O4Os
VUVGYHUDAICLFK-UHFFFAOYSA-N
30318
O=[Os](=O)(=O)=O

Specifications

20816-12-0
Yellow solid
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
O=[Os](=O)(=O)=O
254.226
254.19
Stinging
10 hPa at 20°C
Osmium(VIII) oxide
O4Os
UN 2471
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
Solid
Glass ampoule
1g
Safety and Handling

Safety and Handling

H300 + H310 + H330: Fatal if swallowed, in contact with skin or if inhaled
H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 6.1, Packaging Group: I

EINECSNumber : 244-058-7