missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Osmium(VIII) oxide, 99.8% (Metals basis), Os 74.4% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1210348

 View more versions of this product

Catalog No. AA1210348



Specifications

Specifications

Osmium(VIII) oxide
99.8% (metals basis)
MFCD00011150
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
O=[Os](=O)(=O)=O
254.226
254.1
Like chlorine
10 x 1g
20816-12-0
O4Os
Crystalline
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
Crystalline
10 hPa (8mm Hg) at 20°C

Chemical Identifiers

20816-12-0
254.226
VUVGYHUDAICLFK-UHFFFAOYSA-N
30318
O=[Os](=O)(=O)=O
O4Os
MFCD00011150
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
tetraoxoosmium
Safety and Handling

Safety and Handling

Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H300-H310-H330-H314: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 6.1, Packaging Group: I

EINECSNumber : 244-058-7

Documents