missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Osmium(VIII) oxide, 4% aq. solution, Alfa Aesar™

Manufacturer:  Alfa Aesar 45385AB

 View more versions of this product

Catalog No. AA45385AB



Specifications

Specifications

Osmium(VIII) oxide
O4Os
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
O=[Os](=O)(=O)=O
254.226
254.1
17mm Hg at 20°C
20816-12-0
MFCD00011150
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
Liquid
10mL

Chemical Identifiers

20816-12-0
254.226
VUVGYHUDAICLFK-UHFFFAOYSA-N
30318
O=[Os](=O)(=O)=O
O4Os
MFCD00011150
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
tetraoxoosmium
Safety and Handling

Safety and Handling

H301-H310-H332-H315-H318: Toxic if swallowed. Fatal in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye damage.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 244-058-7

SDS Documents