missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Osmium(VIII) oxide, 2% aq. soln.

Manufacturer:  Alfa Aesar™ 45384AB

 View more versions of this product

Catalog No. AA45384AB



Specifications

Specifications

Osmium (VIII) oxide
O4Os
UN3287
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
O=[Os](=O)(=O)=O
254.226
254.1
2% aq. soln.
Colorless to Yellow
20816-12-0
MFCD00011150
14,6893
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
Liquid
Chlorine-like
10mL

Chemical Identifiers

20816-12-0
254.226
VUVGYHUDAICLFK-UHFFFAOYSA-N
30318
O=[Os](=O)(=O)=O
O4Os
MFCD00011150
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
tetraoxoosmium
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P361-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC LIQUID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents