Learn More
Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 191182500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagentSpecifications
130.0°C | |
40.0°C to 42.0°C | |
Colorless to Yellow | |
250 mg | |
O4Os | |
MFCD00011150 | |
15, 6990 | |
Passes test | |
O=[Os](=O)(=O)=O | |
254.23 | |
254.2 | |
Trace Metal Basis |
Osmium tetroxide | |
20816-12-0 | |
Crystals | |
99.9+% | |
OsO4 | |
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
tetraoxoosmium | |
30318 | |
≥99.9% | |
Ampoule |
Chemical Identifiers
20816-12-0 | |
254.23 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
30318 | |
O=[Os](=O)(=O)=O |
O4Os | |
MFCD00011150 | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
tetraoxoosmium |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
Rinse mouth.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/f
GHS Signal Word: Danger
EINECSNumber : 244-058-7
RUO – Research Use Only