missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, ACROS Organics™

Manufacturer:  Acros Organics AC197450010

 View more versions of this product

Catalog No. AC197450010



Description

Description

Oxidation reagent
Specifications

Specifications

Osmium tetroxide
O4Os
MFCD00011150
15, 6990
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
Fused Mass, Crystals or Lemon Solution
0.8110g/mL
0.811
White to Yellow
4°C
20816-12-0,75-65-0
OsO4
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236
osmic acid
O=[Os](=O)(=O)=O
254.226
254.2
2.5 wt% Min.
Glass bottle
tert-butyl hydroperoxide
1mL

Chemical Identifiers

20816-12-0
254.226
VUVGYHUDAICLFK-UHFFFAOYSA-N
30318
O=[Os](=O)(=O)=O
O4Os
MFCD00011150
osmic acid
tetraoxoosmium
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled. Harmful if swallowed. Fatal in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause drowsiness or dizziness. Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wash face, hands and any exposed skin thoroughly after handling. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger

EINECSNumber : 244-058-7

RTECSNumber : RN1140000

TSCA : TSCA

SDS Documents