Learn More
Osmium(IV) oxide, Os 83% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03949703
Description
The osmium oxide/hexacyanoruthenate films modified electrodes plays an important role for the electrocatalytic reaction of catecholamines and sulfur oxoanions. Used as catalyst. Metallic osmium or an osmium oxide-ruthenium cyanide film modified graphite electrodes plays an essential role for the determination of biogenic amines from electrocatalytic responses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Osmium(IV) oxide | |
12036-02-1 | |
1 g | |
MFCD00011150 | |
Slightly soluble in cold water. | |
O=[Os]=O | |
222.228 | |
222.2 | |
11.4 |
Black | |
Solid | |
O2Os | |
osmium iv oxide, osmium dioxide, osmium oxide oso2 6ci,7ci,8ci,9ci, osmium iv-oxid, acmc-1btxg, osmium iv oxide, os | |
XSXHWVKGUXMUQE-UHFFFAOYSA-N | |
dioxoosmium | |
187574 | |
Black powder |
Chemical Identifiers
12036-02-1 | |
222.228 | |
XSXHWVKGUXMUQE-UHFFFAOYSA-N | |
187574 | |
O=[Os]=O |
O2Os | |
MFCD00011150 | |
osmium iv oxide, osmium dioxide, osmium oxide oso2 6ci,7ci,8ci,9ci, osmium iv-oxid, acmc-1btxg, osmium iv oxide, os | |
dioxoosmium |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only