missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Osmium ICP Standard, 1mL = 1mg Os (1,000ppm Os)OsCl3 in 20% HCl, Ricca Chemical

$308.38 - $308.38

Chemical Identifiers

CAS 7732-18-5,7647-01-0,14996-60-2
CAS Min % 79.0,18.68,0.15
CAS Max % 82.0,19.45,0.15
Physical Form Liquid
Chemical Name or Material Osmium ICP Standard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
POS1KH100 Ricca Chemical Company
RIPOS1KH100
Poly natural 100mL Each for $308.38
POS1KH500 Ricca Chemical Company
RIPOS1KH500
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7647-01-0,14996-60-2
82.0,19.45,0.15
Osmium ICP Standard
79.0,18.68,0.15
Liquid

Specifications

7732-18-5,7647-01-0,14996-60-2
82.0,19.45,0.15
1mL = 1mg Os (1, 000ppm Os)OsCl3 in 20% HCl
Clear black
Osmium ICP Standard
79.0,18.68,0.15
Liquid
Poly natural
100mL

Safety and Handling

ShelfLife : 18 months