missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Osmic Acid Anhydride (Crystalline/Laboratory), Fisher Chemical™
Safety seal, prescored neck, easy-to-snap-open ampules
$493.00 - $493.00
Chemical Identifiers
CAS | 20816-12-0 |
---|---|
Molecular Formula | O4Os |
Molecular Weight (g/mol) | 254.23 |
MDL Number | MFCD00011150 |
InChI Key | VUVGYHUDAICLFK-UHFFFAOYSA-N |
Synonym | osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 |
PubChem CID | 30318 |
IUPAC Name | tetraoxoosmium |
SMILES | O=[Os](=O)(=O)=O |
Chemical Identifiers
20816-12-0 | |
254.23 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
30318 | |
O=[Os](=O)(=O)=O |
O4Os | |
MFCD00011150 | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
tetraoxoosmium |
Specifications
130°C | |
White | |
Solid | |
O4Os | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
O=[Os](=O)(=O)=O | |
254.23 | |
254.23 | |
Pass Test | |
7mmHg at 20°C | |
Osmic Acid Anhydride |
39.5°C | |
20816-12-0 | |
0.25 g | |
MFCD00011150 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
tetraoxoosmium | |
30318 | |
Laboratory | |
Ampule | |
4.9g/cm³ |
Safety and Handling
DOTInformation : DOT Class 6.1, : Poison