Osmette II™ and Osmette III™ Osmometers
The simplicity and dependability of Osmette™ osmometers in a small, lightweight size. Osmette II and Osmette III Osmometers are ideal for one-step freezing-point measurement of osmolality.
Manufacturer:  Precision Systems Inc PSY5005
 View more versions of this product
Catalog No. 12-827-31
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
≈2 to 3 min.
Each
Osmette™ ii Osmometer
8.5 in.
35.6cm
10μL
Pediatric, neonatal, geriatric and other applications
50/60Hz
0.5A
14 x 8.5 x 5 in. (35.6 x 21.6 x 12.7cm)
5 in.
9 lb.
12.7cm
5005
4.1kg
14 in.
21.6cm
0 to 2000mOsm/kg H2O
115V
115V 50/60Hz
Auto operation
60 to 90 sec.