Please login to your online account to display your discounted pricing

OSM™ All-Natural Glass and Surface Cleaner

Streak-free shine with no smearing

$230.74 - $235.54

Specifications

Boiling Point >100°C
Color Clear
For Use With (Application) Use in lab, on lab Glassware, kitchen, bathroom or wherever Glass is present
Format Liquid
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

15336503

 
organic sales & mktg inc
81008201237
32 oz. Case of 12 Each for $230.74

15336512

 
organic sales & mktg inc
81008201238
1 gal. Case of 4 Each for $235.54
Description & Specifications

Specifications

Boiling Point >100°C
Color Clear
For Use With (Application) Use in lab, on lab Glassware, kitchen, bathroom or wherever Glass is present
Format Liquid

Ideal for a variety of surfaces in the bathroom, kitchen, and work areas. Use on glass, plastic, rubber, vinyl, labware, and most metals. Renewable plant-derived formula free of phosphates, glycol ether, phthalates, and ammonia.

  • Perfect for glass test tubes, Petri dishes, and weigh plates
  • Water-based product with natural cleaning agents and plant-based surfactants
  • Environmentally friendly alternative to harsh chemicals; safe to use around people and animals
  • Disposes down drain
  • No ammonia, no harsh chemical fumes or vapors

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.