Please login to your online account to display your discounted pricing

OSM™ All-Natural Degreaser

Cuts through grease and grime in seconds

$229.40 - $229.40

Specifications

Certifications/Compliance 29 CFR1910.1200
Color Clear Light Blue
For Use With (Application) Ideal for steam cleaning engines and machinery or for general cleanups in and around facilities
Format Liquid
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

15336511

 
organic sales & mktg inc
81008201228
1 gal. Case of 4 Each for $229.40
Description & Specifications

Specifications

Certifications/Compliance 29 CFR1910.1200
Color Clear Light Blue
For Use With (Application) Ideal for steam cleaning engines and machinery or for general cleanups in and around facilities
Format Liquid

Ideal for use on all lab and office surfaces.

  • Removes grease and oil naturally
  • Plant-based cleaning agents contain no petroleum solvents
  • Safe to use around people and animals
  • Ozone-safe formula disposes down the drain
  • No harsh chemical fumes or vapors

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.