Please login to your online account to display your discounted pricing

OSM™ All-Natural All-Purpose Cleaner

Powerful cleaning action

$17.55 - $17.55

Specifications

Boiling Point >100°C
Color Clear Amber
For Use With (Application) Ideal for use on countertops, floors, cabinets, appliances and all lab surfaces
Format Liquid
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

S95747

 
fisher scientific
81008201207/EMD
32 oz. Each for $17.55
Case of 12 Each for $191.70
Description & Specifications

Specifications

Boiling Point >100°C
Color Clear Amber
For Use With (Application) Ideal for use on countertops, floors, cabinets, appliances and all lab surfaces
Format Liquid

  • Ideal for use on countertops, floors, cabinets, appliances, and all lab surfaces
  • Perfect for use in healthcare, vivariums, and animal environments
  • Penetrates and emulsifies dirt, grease, and grime
  • Enzyme-based cleaning agents with plant-based surfactants
  • Noncaustic, nontoxic, nonallergenic, and biodegradable
  • Renewable, plant-derived formula free of phosphates, glycol ether, and phthalate
  • Safe to use around people and pets; disposes down drain
  • No harsh chemical fumes or vapors
  • Ozone safe and environmentally preferable