Please login to your online account to display your discounted pricing

OSM™ All-Natural All-Purpose Cleaner

Powerful cleaning action

$214.11 - $228.14

Specifications

Boiling Point >100°C
Color Clear Amber
For Use With (Application) Ideal for use in retail, industrial, school and office environments, Ideal for use on countertops, floors, cabinets, appliances and all lab surfaces
Format Liquid
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

15336500

 
organic sales & mktg inc
81008201207
32 oz. Case of 12 Each for $228.14

15336509

 
organic sales & mktg inc
81008201208
1 gal. Case of 4 Each for $214.11
Description & Specifications

Specifications

Boiling Point >100°C
Color Clear Amber
For Use With (Application) Ideal for use in retail, industrial, school and office environments, Ideal for use on countertops, floors, cabinets, appliances and all lab surfaces
Format Liquid

Ideal for use on countertops, floors, cabinets, appliances, and all lab surfaces. Renewable, plant-derived formula free of phosphates, glycol ether, and phthalate.

  • Perfect for use in healthcare and animal environments
  • Penetrates and emulsifies dirt, grease, and grime
  • Plant-based surfactants and cleaning agents
  • Safe to use around people and pets; disposes down drain
  • No harsh chemical fumes or vapors
  • Ozone safe and environmentally preferable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.