Please login to your online account to display your discounted pricing

OSM™ All-Natural All-Purpose Cleaner


Powerful cleaning action

Manufacturer: fisher scientific  81008201207/EMD

 View more versions of this product

Catalog No. S95747

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Color Clear Amber
Format Liquid
Boiling Point >100°C
For Use With (Application) Ideal for use on countertops, floors, cabinets, appliances and all lab surfaces
Size 32 oz.