Please login to your online account to display your discounted pricing

Orthophenanthroline TS, 0.025M Phenanthroline Ferrous Sulfate, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Orthophenanthroline TS
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 5144-89-8
CAS Min % 1.44
CAS Max % 1.5
CAS 7782-63-0
CAS Min % 0.67
CAS Max % 0.7
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

54648-16

 
ricca chemical company
RI5464.816
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

54648-4

 
ricca chemical company
RI5464.84
Glass amber 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear dark red
Quantity 500mL
Chemical Name or Material Orthophenanthroline TS
CAS 7732-18-5
CAS Max % 100.0
CAS 7782-63-0
Shelf Life 24 months
CAS 5144-89-8
CAS Min % 96.0
Concentration or Composition (by Analyte or Components) 0.025M Phenanthroline Ferrous Sulfate
CAS Min % 0.67
CAS Min % 1.44
CAS Max % 0.7
Packaging Glass amber
CAS Max % 1.5
Safety and Handling

24 months