Ortho-Clinical Diagnostics™ ID-Micro Typing System™ (MTS) Instrumentation DFS Item
Suitable for a broad range of blood bank applications. These accessories are used with the Ortho Workstation (OK6904630).
$2,291.82 - $2,291.82
Specifications
For Use With (Equipment) Ortho™ Workstation
View More Specs
Requires: ID-MTS Manual GEL Workstation™, ORTHO ProVue™
Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
OKMTS9640 DFS Item
View Documents
Ortho-Clinical Diagnostics™
MTS9640
ID-TipMaster™ Pipettor
Each for $2,291.82
Description
Description
ID-Micro Typing System™ (ID-MTS)
  • Improved sensitivity and specificity
  • No-wash antiglobulin procedure
  • Standardized procedures
  • Improved turnaround time
  • Enhanced regulatory compliance
Antibody screening and identification, ABO blood grouping and Rh phenotyping, Compatibility Testing, Reverse serum grouping, Antigen typing
Specifications
Specifications
Ortho™ Workstation
SDS