Ortho-Clinical Diagnostics™ ID-Micro Typing System™ (MTS) Instrumentation
Suitable for a broad range of blood bank applications. These accessories are used with the Ortho Workstation (OK6904630).
Manufacturer: Ortho-Clinical Diagnostics™ MTS9640
Requires : ID-MTS Manual GEL Workstation™, ORTHO ProVue™
Description
ID-Micro Typing System™ (ID-MTS)
- Improved sensitivity and specificity
- No-wash antiglobulin procedure
- Standardized procedures
- Improved turnaround time
- Enhanced regulatory compliance
Antibody screening and identification, ABO blood grouping and Rh phenotyping, Compatibility Testing, Reverse serum grouping, Antigen typing
Specifications
Ortho™ Workstation | |
ID-TipMaster™ Pipettor |