Please login to your online account to display your discounted pricing

Alfa Aesar™ Orthanilic acid, 97%

$85.89 - $85.89

Chemical Identifiers

Chemical Name or Material Orthanilic acid
CAS 88-21-1
Molecular Formula C6H7NO3S
Formula Weight 173.19
MDL Number MFCD00007705
Synonym Aniline-2-sulfonic acid; 2-Aminobenzenesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3176322

  SDS
alfa aesar™
H3176322
100g Each for $85.89
Description & Specifications

Specifications

Formula Weight 173.19
Chemical Name or Material Orthanilic acid
TSCA TSCA
Assay 97%
Molecular Formula C6H7NO3S
MDL Number MFCD00007705
CAS 88-21-1
Synonym Aniline-2-sulfonic acid; 2-Aminobenzenesulfonic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point >300°C
Odor Odorless
Quantity 100g
RTECS Number DB4727700
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.