Please login to your online account to display your discounted pricing

ORP Standard, 475mV, Certified, LabChem™

$78.06 - $78.06

Chemical Identifiers

Chemical Name or Material ORP Standard
CAS 7732-18-5
CAS Max % 81.76
CAS 7664-93-9
CAS Max % 9.5
CAS 7783-83-7
CAS Max % 4.82
CAS 7783-85-9
CAS Max % 3.92
Name Note 475mV
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC180201

 
lab chem
LC180201
Amber Poly 500mL Each for $78.06
Description & Specifications

Specifications

Formula Weight 392.14
Purity Grade Certified Grade
UN Number UN3264
CAS Max % 9.5
Name Note 475mV
Identification Passes Test
Chemical Name or Material ORP Standard
CAS Max % 4.82
Solubility Soluble in water
Quantity 500mL
Linear Formula Fe(NH4)2(SO4)2·6H2O
CAS 7783-83-7
Health Hazard 1 Danger
Synonym Light's Solution
CAS Max % 3.92
CAS 7783-85-9
CAS 7732-18-5
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 81.76
Packaging Amber Poly
Physical Form Liquid
Recommended Storage Room Temperature
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Color Clear, dull yellow
CAS 7664-93-9
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.

Room Temperature