Please login to your online account to display your discounted pricing

ORP Standard, 400mV, Certified, LabChem™

$73.17 - $73.17

Chemical Identifiers

Chemical Name or Material ORP Standard
CAS 7732-18-5
CAS Max % 88.1
CAS 7783-85-9
CAS Max % 9.1
CAS 7664-93-9
CAS Max % 1.8
CAS 7783-83-7
CAS Max % 1
Name Note 400mV
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC180151

 
lab chem
LC180151
Amber Poly 500mL Each for $73.17
Description & Specifications

Specifications

Chemical Name or Material ORP Standard
Name Note 400mV
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 88.1
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 7783-85-9
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 9.1
Recommended Storage Room Temperature
CAS 7664-93-9
CAS Max % 1.8
CAS 7783-83-7
CAS Max % 1
Purity Grade Certified Grade
Physical Form Liquid
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.14
UN Number UN2796
Identification Passes Test
Packaging Amber Poly
Solubility Soluble in water
Density 1.06g/mL
Color Clear, dull yellow
Quantity 500mL
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.

Room Temperature