Please login to your online account to display your discounted pricing

ORP Standard, 200mV, Certified, LabChem™

$67.00 - $67.00

Chemical Identifiers

Chemical Name or Material ORP Standard
CAS 7732-18-5
CAS Max % 98.64
CAS 14459-95-1
CAS Max % 0.53
CAS 7447-40-7
CAS Max % 0.75
CAS 13746-66-2
CAS Max % 0.08
Name Note 200mV
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC180101

 
lab chem
LC180101
Amber Poly 500mL Each for $67.00
Description & Specifications

Specifications

Chemical Name or Material ORP Standard
Name Note 200mV
CAS 7732-18-5
CAS Max % 98.64
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 14459-95-1
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.53
Recommended Storage Room Temperature
CAS 7447-40-7
CAS Max % 0.75
CAS 13746-66-2
CAS Max % 0.08
Purity Grade Certified Grade
Physical Form Liquid
Linear Formula K3Fe(CN)6
Formula Weight 329.26
Identification Passes Test
Packaging Amber Poly
Solubility Soluble in water
Density 1g/mL
Color Clear, yellow
Quantity 500mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature