missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Orotic Acid Monohydrate 98.0+%, TCI America™
Supplier: TCI America O0065500G
Specifications
Orotic Acid Monohydrate | |
White | |
MFCD00006027 | |
orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate | |
OC(=O)C1=CC(=O)NC(=O)N1 | |
156.10 | |
156.10 | |
Crystalline Powder |
50887-69-9 | |
C5H4N2O4 | |
500 g | |
PXQPEWDEAKTCGB-UHFFFAOYSA-N | |
2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid | |
16218504 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
50887-69-9 | |
156.10 | |
PXQPEWDEAKTCGB-UHFFFAOYSA-N | |
16218504 | |
OC(=O)C1=CC(=O)NC(=O)N1 |
C5H4N2O4 | |
MFCD00006027 | |
orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate | |
2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid |
Safety and Handling
EINECSNumber : (9)-0278
RTECSNumber : RM3180000
TSCA : Yes